Please login to your online account to display your discounted pricing

Sino Biological™ mMP8 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the pro form of humanmMP8 (NP_002415.1) (Phe 21-Gly 467) fused to an initial Met at the N-terminus

$1,208.00 - $1,208.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias MMP8, CLG1, HNC, mMP-8, PMNL-CL
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10254HNAH50

DFS Item
 
life technologies
LS10254HNAH50
50μg Each for $1,208.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias MMP8, CLG1, HNC, mMP-8, PMNL-CL

Matrix Metallopeptidase 8 (MMP8), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate, Mca-PLGL-Dpa-AR-NH2 (R&D Systems, Catat. No. ES001), the specific activity is >250pmoles/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.