Please login to your online account to display your discounted pricing

Sino Biological™ mMP8 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the humanmMP8 (NP_002415.1) (Met 1-Gly 467) fused to the polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10254H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10254H08H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 52.6kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purity ≥85% by SDS-PAGE
Validated Application Assay Standard
Expression System Mammalian
Common Name Matrix metallopeptidase 8 (neutrophil collagenase)
Protein Subtype Proteases
Protein Family Other Proteins
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias CLG1, HNC, PMNL-CL, mMP8
Endotoxin Concentration <1 EU/μg
Gene Symbol MMP8
Protein Tag His Tag
Format Lyophilized

Matrix metallopeptidase 8 (neutrophil collagenase) (MMP8), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate, Mca-PLGL-Dpa-AR-NH2, the specific activity is >450pmoles/min./μg
  • Formulation: Lyophilized in 50mM Tris, 10mM CaCl2, 150mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.