missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ mMP8 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the pro form of humanmMP8 (NP_002415.1) (Phe 21-Gly 467) fused to an initial Met at the N-terminus

Manufacturer:  Life Technologies LS10254HNAH50

 View more versions of this product

Catalog No. 10254HNAH50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological;mMP8 Recombinant Human Protein; 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Matrix Metallopeptidase 8 (MMP8), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate, Mca-PLGL-Dpa-AR-NH2 (R&D Systems, Catat. No. ES001), the specific activity is >250pmoles/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
52kDa
50μg
Proteases
MMP8, CLG1, HNC, mMP-8, PMNL-CL
MMP8
Other Proteins
Untagged
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Matrix metallopeptidase 8
Assay Standard
Mammalian