Please login to your online account to display your discounted pricing

Sino Biological™ mMP7 Recombinant Human Protein, N-hIgG1-Fc Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the active form of humanmMP7 (NP_002414.1) (Tyr 95-Lys 267) fused to Fc region of human IgG1 at the N-terminus

Manufacturer: life technologies  LS10277H01H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10277H01H25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 45.8kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination N-Terminal
Structural Form Full Length/Recombinant
Protein Subtype Proteases
Endotoxin Concentration <1 EU/μg
Gene Alias MMP7, mMP-7, MPSL1, PUMP-1
Common Name Matrix metallopeptidase 7
Gene Symbol MMP7
Protein Tag IgG1 Fc Tag
Validated Application Assay Standard
Protein Family Proteases, Peptidases & Inhibitors
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 4316

Matrix Metallopeptidase 7 (MMP7), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM Glycine, 10mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.