missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ mMP3 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the humanmMP3 (AAA36321.1) N-terminal fragment (Tyr 18-Thr 272)

$174.00 - $762.00

Specifications

Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Alias CHDS6, mMP-3, SL-1, STMY, STMY1, STR1
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10467HNAE25 Life Technologies
LS10467HNAE25
5 x 5μg Each for $476.00
10467HNAE50 Life Technologies
LS10467HNAE50
50μg Each for $762.00
10467HNAE5 Life Technologies
LS10467HNAE5
5μg Each for $174.00
Specifications

Description

Matrix Metallopeptidase 3 (MMP3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (AnaSpec Cat. No. 27114), the specific activity is >300pmoles/min./μg
  • Formulation: Lyophilized in 50mM Tris, 10mMCaCl2, 1μM ZnCl2, 50mM NaCl, 0.5%Brij35 pH 7.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
CHDS6, mMP-3, SL-1, STMY, STMY1, STR1