missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ mMP3 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the humanmMP3 (AAA36321.1) N-terminal fragment (Tyr 18-Thr 272)

Manufacturer:  Life Technologies LS10467HNAE50

 View more versions of this product

Catalog No. 10467HNAE50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological;mMP3 Recombinant Human Protein; 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Matrix Metallopeptidase 3 (MMP3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate, Mca-RPKPVE-Nval-WRK(Dnp)-NH2 (AnaSpec Cat. No. 27114), the specific activity is >300pmoles/min./μg
  • Formulation: Lyophilized in 50mM Tris, 10mMCaCl2, 1μM ZnCl2, 50mM NaCl, 0.5%Brij35 pH 7.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
29kDa
50μg
Proteases
Matrix metallopeptidase 3
Assay Standard
E. coli
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
CHDS6, mMP-3, SL-1, STMY, STMY1, STR1
MMP3
Other Proteins
Untagged