missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ mMP1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the pro form of humanmMP1 enzyme (NP_002412.1) precursor (Met 1-Asn 469) fused to a C-terminal polyhistidine tag

Manufacturer:  Life Technologies LS10532H08H25

 View more versions of this product

Catalog No. 10532H08H25


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological;mMP1 Recombinant Human Protein, His Tag; 5 x 5ug; Source: Human; Purity: greater than or equal to 95% by SDS-PAGE

Matrix Metallopeptidase 1 (MMP1), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate, Mca-PLGL-Dpa-AR-NH2 (AnaSpec Cat. No. 27076), the specific activity is >70pmole/min./μg
  • Formulation: 25mM MES, 10mM CaCl2, 150mM NaCl, 0.05% BRIG 35, pH 5.5
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
52kDa
5 x 5μg
Proteases
CLG, CLGN
MMP1
Other Proteins
His Tag
Frozen
≥95% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Matrix Metallopeptidase 1
Assay Standard
Mammalian