missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MIF Recombinant Mouse Protein, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse MIF (P34884) precursor (Pro 2-Ala 115) fused to an initial Met at the C-terminus

$99.75 - $429.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias GIF, Glif, mgC107654
View More Specs

Includes: Five vials of protein in suspension buffer

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50066MNAE5 View Documents Life Technologies
LS50066MNAE5
5μg Each for $99.75
 
50066MNAE50 View Documents Life Technologies
LS50066MNAE50
50μg Each for $429.00
 
Description

Description

Macrophage Migration Inhibitory Factor (MIF), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 8.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
GIF, Glif, mgC107654
PAGE
Lyophilized
17319

For Research Use Only. Not for use in diagnostic procedures.