missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MIF Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human MIF (P14174) (Pro 2-Ala 115) fused to a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus

$196.00 - $530.00

Specifications

Purity ≥90% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias GIF, GLIF, mMIF
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10246H08H25 View Documents Life Technologies
LS10246H08H25
5 x 5μg Each for $530.00
 
10246H08H5 View Documents Life Technologies
LS10246H08H5
5μg Each for $196.00
 
Description

Description

Macrophage Migration Inhibitory Factor (MIF), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
GIF, GLIF, mMIF
PAGE
Lyophilized
4282

For Research Use Only. Not for use in diagnostic procedures.