Please login to your online account to display your discounted pricing

Sino Biological™ MICB Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human MICB (NP_005922.2) (Met 1-Gly 298) fused to a C-terminal polyhistidine tag

Manufacturer: life technologies  LS10759H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10759H08H50

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 4277
Molecular Weight 33kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Endotoxin Concentration <1 EU/μg
Termination C-Terminal
Format Lyophilized
Gene Alias DAAP-210H10.1, PERB11.2, MICB
Purity ≥90% by SDS-PAGE
Structural Form Full Length/Recombinant
Validated Application Assay Standard
Common Name MHC Class I polypeptide-related sequence B
Protein Family MHC (Major Histocompatibility Complex) Proteins
Protein Tag His Tag
Gene Symbol MICB
Purification Method PAGE
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Expression System Mammalian
Quantity 50μg

MHC Class I Polypeptide-Related Sequence B (MICB), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.