Please login to your online account to display your discounted pricing

Sino Biological™ MEP1A Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse MEP1A (NP_032611.2) (Met 1-Arg 615) fused with a polyhistidine tag at the C-terminus

$221.00 - $3,845.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AI098089, AW107200, Mep-1, Mep-1a, Mep1
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50057M08H25

DFS Item
 
life technologies
LS50057M08H25
5 x 5μg Each for $700.00

50057M08H5

DFS Item
 
life technologies
LS50057M08H5
5μg Each for $221.00

50057M08H50

DFS Item
 
life technologies
LS50057M08H50
50μg Each for $1,124.00

50057M08HL

DFS Item
 
life technologies
LS50057M08H250
5 x 50μg Each for $3,845.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias AI098089, AW107200, Mep-1, Mep-1a, Mep1

Meprin 1 alpha (MEP1A), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to cleave a fluorogenic peptide substrate, Mca-YVADAPK(Dnp)-OH; the specific activity is >400pmole/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.