Please login to your online account to display your discounted pricing

Sino Biological™ MCAM Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human MCAM precursor (NP_006491.2) (Met 1-Gly 559) fused to a C-terminal polyhistidine tag

Manufacturer: life technologies  LS10115H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10115H08H25

  • / Each
 

Description & Specifications

Specifications

Validated Application Assay Standard
Quantity 5 x 5μg
Endotoxin Concentration <1 EU/μg
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.Expires one year from date of receipt when stored as instructed.
Purity ≥90% by SDS-PAGE
Format Lyophilized
Gene Symbol MCAM
Common Name Melanoma Cell Adhesion Molecule
Protein Tag His Tag
Gene Alias MCAM, CD146, MUC18
Protein Subtype Intercellular Adhesion Molecule
Protein Family CD Proteins
Molecular Weight 61.3kDa
Expression System Mammalian

Melanoma Cell Adhesion Molecule (MCAM), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by the ability of the immobilized protein to support the adhesion of the HUVEC human umbilical vein endothelial cell line; when 3 x 104 cells/well are added to human MCAM coated plates (0.8μg/mL, 100μL/well), approximately >50% will adhere after one hour at 37°C
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.