missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

MCAM (CD146) Recombinant Human Protein, hIgG1-Fc Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 559) of human MCAM precursor (NP_006491.2) fused to the Fc region of human IgG1 at the C-terminus

$606.00 - $606.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias MCAM, CD146, MUC18
Gene ID (Entrez) 4162
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10115H02H50 View Documents Life Technologies
LS10115H02H50
50μg Each for $606.00
 
Description

Description

Melanoma cell adhesion molecule (MCAM/CD146), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in cell biology research applications. It can also be used as a protein standard.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of the HUVEC human umbilical vein endothelial cell line; when 3 x 104 cells/well are added to human CD146 coated plates (0.8μg/mL, 100μL/well), approximately >30 % will adhere after one hour at 37°C
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
4162
<1 EU/μg
MCAM, CD146, MUC18

For Research Use Only. Not for use in diagnostic procedures.