Please login to your online account to display your discounted pricing

Sino Biological™ MAP1D Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human METAP1D (NP_954697.1) (Arg 44-Ala 335) with a polyhistidine tag at the C-terminus and an initial Met an the N-terminus

$159.00 - $2,530.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias MAP1D
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10883H08B25

DFS Item
 
life technologies
LS10883H08B25
5 x 5μg Each for $475.00

10883H08B5

DFS Item
 
life technologies
LS10883H08B5
5μg Each for $159.00

10883H08B50

DFS Item
 
life technologies
LS10883H08B50
50μg Each for $720.00

10883H08BL

DFS Item
 
life technologies
LS10883H08B250
5 x 50μg Each for $2,530.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias MAP1D

Methionyl aminopeptidase type 1D (mitochondrial) (METAP1D), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measure by its ability to remove methionine from a fluorogenic peptide substrate H-Met-Gly-Pro-AMC; the resulting Gly-Pro-AMC is cleaved by human DPP4/CD26, the specific activity is >30pmole/min./μg
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.