missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

LUM Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human LUM (NP_002336.1) (Met 1-Asn 338) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Invitrogen LS11640H08H50

 View more versions of this product

Catalog No. 11640H08H50



Description

Description

Lumican (LUM), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Mammalian
Assay Standard
Lyophilized
38kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
LDC, SLRR2D
LUM
Cell Structure/Organelle Proteins
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
4060
PAGE
50μg
C-Terminal
<1 EU/μg
Lumican
His Tag
SDS
Documents

For Research Use Only. Not for use in diagnostic procedures.