Please login to your online account to display your discounted pricing

Sino Biological™ LTA Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human LTA (P01374) (Leu 35-Leu 205) fused to an initial Met at the N-terminus

$106.00 - $1,320.00

Specifications

Purity ≥95% by SDS-PAGE
Format Lyophilized
Gene Alias LTA, DAMA-25N12, 13-004, LT, TNFB, TNFSF1
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10270HNAE5

DFS Item
 
life technologies
LS10270HNAE5
5μg Each for $106.00

10270HNAE25

DFS Item
 
life technologies
LS10270HNAE25
5 x 5μg Each for $308.00

10270HNAEL

DFS Item
 
life technologies
LS10270HNAE250
5 x 50μg Each for $1,320.00
Description & Specifications

Specifications

Purity ≥95% by SDS-PAGE
Format Lyophilized
Gene Alias LTA, DAMA-25N12, 13-004, LT, TNFB, TNFSF1

Lymphotoxin Alpha (TNF Superfamily Member 1) (LTA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured in a cytotoxicity assay using L929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D, the ED50 for this effect is typically 0.1-0.4ng/mL
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.