Please login to your online account to display your discounted pricing

Sino Biological™ LTA Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the mature form of human LTA (P01374) (Leu 35-Leu 205) fused to an initial Met at the N-terminus

Manufacturer: life technologies  LS10270HNAE25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10270HNAE25

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 18.8kDa
Purity ≥95% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype TNF (Tumor Necrosis Factor)
Gene Alias LTA, DAMA-25N12, 13-004, LT, TNFB, TNFSF1
Common Name Lymphotoxin alpha (TNF superfamily, member 1)
Gene Symbol LTA
Protein Tag Untagged
Validated Application Assay Standard
Protein Family Cytokines & Receptors
Expression System E. coli
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Lymphotoxin Alpha (TNF Superfamily Member 1) (LTA), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured in a cytotoxicity assay using L929 mouse fibrosarcoma cells in the presence of the metabolic inhibitor actinomycin D, the ED50 for this effect is typically 0.1-0.4ng/mL
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.