Please login to your online account to display your discounted pricing

Sino Biological™ LRPAP1 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human LRPAP1 (NP_002328.1) (Tyr 35-Leu 357) fused to the signal peptide at the N-terminus and a polyhistidine tag at the C-terminus

$97.50 - $1,320.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias LRPAP1, RP11-529E10.3, A2MRAP, A2RAP, HBP44, mgC138272, MRAP, RAP
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

11100H08H25

DFS Item
 
life technologies
LS11100H08H25
5 x 5μg Each for $218.00

11100H08H5

DFS Item
 
life technologies
LS11100H08H5
5μg Each for $97.50

11100H08HL

DFS Item
 
life technologies
LS11100H08H250
5 x 50μg Each for $1,320.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias LRPAP1, RP11-529E10.3, A2MRAP, A2RAP, HBP44, mgC138272, MRAP, RAP

Low density lipoprotein receptor-related protein associated protein 1 (LRPAP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human LRPAP1 at 0.5μg/mL can bind human VLDLR with a range of 3.2-400ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.