Please login to your online account to display your discounted pricing

Sino Biological™ LRPAP1 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human LRPAP1 (NP_002328.1) (Tyr 35-Leu 357) fused to the signal peptide at the N-terminus and a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11100H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11100H08H5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 39.2kDa
Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias LRPAP1, RP11-529E10.3, A2MRAP, A2RAP, HBP44, mgC138272, MRAP, RAP
Common Name Low density lipoprotein receptor-related protein associated protein 1
Gene Symbol LRPAP1
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Low density lipoprotein receptor-related protein associated protein 1 (LRPAP1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human LRPAP1 at 0.5μg/mL can bind human VLDLR with a range of 3.2-400ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.