missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

LGALS8 Recombinant Human Protein, N-GST Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding human LGALS8 (AAF19370.1) (Met 1-Trp 317) fused to a GST tag at the N-terminus

$226.00 - $226.00

Specifications

Purity >76% by SDS-PAGE
Format Lyophilized
Gene Alias LGALS8, RP11-385F5.1, Gal-8, PCTA-1, PCTA1, Po66-CBP
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Qty  
Catalog Number Mfr. No. Quantity Price Qty  
10301H09E25 missing translation for 'viewDocuments' Life Technologies
LS10301H09E25
5 x 5μg Each for $226.00
 
Description

Description

Lectin, galactoside-binding, soluble, 8 (LGALS8), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its ability to agglutinate human red blood cells, the ED50 for this effect is typically 0.2-1μg/mL
  • Formulation: Lyophilized in 50mM Tris, 0.1M NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

>76% by SDS-PAGE
Lyophilized
LGALS8, RP11-385F5.1, Gal-8, PCTA-1, PCTA1, Po66-CBP

For Research Use Only. Not for use in diagnostic procedures.