Please login to your online account to display your discounted pricing

Sino Biological™ LGALS8 Recombinant Human Protein, N-GST Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding human LGALS8 (AAF19370.1) (Met 1-Trp 317) fused to a GST tag at the N-terminus

Manufacturer: life technologies  LS10301H09E5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10301H09E5

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 62.7kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Purity >76% by SDS-PAGE
Format Lyophilized
Protein Family Other Proteins
Protein Tag GST Tag
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Gene Alias LGALS8, RP11-385F5.1, Gal-8, PCTA-1, PCTA1, Po66-CBP
Expression System E. coli
Quantity 5μg
Protein Subtype Lectins & Leptins
Common Name Lectin, galactoside-binding, soluble, 8
Gene Symbol LGALS8
Validated Application Assay Standard

Lectin, galactoside-binding, soluble, 8 (LGALS8), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by its ability to agglutinate human red blood cells, the ED50 for this effect is typically 0.2-1μg/mL
  • Formulation: Lyophilized in 50mM Tris, 0.1M NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.