Please login to your online account to display your discounted pricing

Sino Biological™ LGALS3 Recombinant Human Protein, Low Endotoxin DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human LGALS3 (P17931) (Met 1-Ile 250)

$241.00 - $3,740.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <5 EU/mg
Format Lyophilized
Gene Alias LGALS3, CBP35, GAL3, GALBP, GALIG, L31, LGALS2, MAC2
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10289HNAEE5

DFS Item
 
life technologies
LS10289HNAEE5
5μg Each for $241.00

10289HNAEES

DFS Item
 
life technologies
LS10289HNAEE50
50μg Each for $1,140.00

10289HNAEEL

DFS Item
 
life technologies
LS10289HNAEE250
5 x 50μg Each for $3,740.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <5 EU/mg
Format Lyophilized
Gene Alias LGALS3, CBP35, GAL3, GALBP, GALIG, L31, LGALS2, MAC2

Lectin, galactoside-binding, soluble, 3 (LGALS3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by its ability to agglutinate human red blood cells, the ED50 for this effect is typically 3-15μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.