missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ LGALS3 Recombinant Human Protein, Low Endotoxin DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human LGALS3 (P17931) (Met 1-Ile 250)

Manufacturer:  Life Technologies LS10289HNAEE250

 View more versions of this product

Catalog No. 10289HNAEEL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; LGALS3 Recombinant Human Protein (Low Endotoxin); 5 x 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Lectin, galactoside-binding, soluble, 3 (LGALS3), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by its ability to agglutinate human red blood cells, the ED50 for this effect is typically 3-15μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
26.1kDa
5 x 50μg
Lectins & Leptins
LGALS3, CBP35, GAL3, GALBP, GALIG, L31, LGALS2, MAC2
LGALS3
Other Proteins
Untagged
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<5 EU/mg
Lectin, galactoside-binding, soluble, 3
Assay Standard
E. coli