Please login to your online account to display your discounted pricing

Sino Biological™ LGALS1 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human LGALS1 (NP_002296.1) (Met 1-Asp135)

$98.25 - $764.00

Specifications

Purity ≥95% by SDS-PAGE
Format Lyophilized
Gene Alias LGALS1, DKFZp686E23103, GAL1, GBP
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10290HNAE25

DFS Item
 
life technologies
LS10290HNAE25
5 x 5μg Each for $216.00

10290HNAE5

DFS Item
 
life technologies
LS10290HNAE5
5μg Each for $98.25

10290HNAEL

DFS Item
 
life technologies
LS10290HNAE250
5 x 50μg Each for $764.00
Description & Specifications

Specifications

Purity ≥95% by SDS-PAGE
Format Lyophilized
Gene Alias LGALS1, DKFZp686E23103, GAL1, GBP

Lectin, galactoside-binding, soluble, 1 (LGALS1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by its ability to agglutinate human red blood cells, the ED50 for this effect is typically 0.5-3μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.