Please login to your online account to display your discounted pricing

Sino Biological™ LGALS1 Recombinant Human Protein DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human LGALS1 (NP_002296.1) (Met 1-Asp135)

Manufacturer: life technologies  LS10290HNAE25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10290HNAE25

  • / Each
 

Description & Specifications

Specifications

Validated Application Assay Standard
Molecular Weight 15kDa
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Expression System E. coli
Protein Family Other Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Common Name Lectin, galactoside-binding, soluble, 1
Gene Symbol LGALS1
Gene Alias LGALS1, DKFZp686E23103, GAL1, GBP
Protein Tag Untagged
Purity ≥95% by SDS-PAGE
Format Lyophilized
Protein Subtype Lectins & Leptins

Lectin, galactoside-binding, soluble, 1 (LGALS1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by its ability to agglutinate human red blood cells, the ED50 for this effect is typically 0.5-3μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.