Please login to your online account to display your discounted pricing

Sino Biological™ LEPR Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Asp 839) of human LEPR (NP_002294.2) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10322H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10322H08H50

  • / Each
 

Description & Specifications

Specifications

Common Name Leptin receptor
Protein Tag His Tag
Gene Symbol LEPR
Purity ≥95% by SDS-PAGE
Quantity 50μg
Protein Subtype Cytokine Receptors|Hormone Receptors
Protein Family Other Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Alias LEPR, CD295, OBR
Expression System Mammalian
Format Lyophilized
Validated Application Assay Standard
Endotoxin Concentration <1 EU/μg
Molecular Weight 95kDa
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.

Leptin receptor (LEPR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human LEPR at 5μg/mL (100μL/well) can bind human leptin receptor (LEPR) with a linear range of 0.032-4.0μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.