missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

LEP (Leptin) Recombinant Human Protein, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human Leptin (NP_000221.1), residues 22 to 167, containing an additional Met

$78.75 - $142.00

Specifications

Purity ≥95% by SDS-PAGE
Format Lyophilized
Gene Alias FLJ94114, OB, OBS
Gene ID (Entrez) 3952
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10221HNAEL View Documents Invitrogen
LS10221HNAE250
5 x 50μg Each for $142.00
 
10221HNAE5 View Documents Invitrogen
LS10221HNAE5
5μg Each for $78.75
 
10221HNAE50 View Documents Invitrogen
LS10221HNAE50
50μg Each for $110.00
 
Description

Description

Leptin (LEP), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human Leptin at 1.25μg/mL (100μL/well) can bind human Leptin receptor-Fc with a linear range of 0.032-4.0μg/mL; 2. Immobilized human Leptin at 5μg/mL (100μL/well) can bind human Leptin receptor-His with a linear range of 0.032-4.0μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥95% by SDS-PAGE
Lyophilized
FLJ94114, OB, OBS
3952

For Research Use Only. Not for use in diagnostic procedures.