missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

LEP (Leptin) Recombinant Human Protein, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human Leptin (NP_000221.1), residues 22 to 167, containing an additional Met

Manufacturer:  Life Technologies LS10221HNAE5

 View more versions of this product

Catalog No. 10221HNAE5



Description

Description

Leptin (LEP), recombinant human protein is supplied as a lyophilized powder. In general, recombinant proteins are suitable for use in studying protein-protein interactions, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA: 1. Immobilized human Leptin at 1.25μg/mL (100μL/well) can bind human Leptin receptor-Fc with a linear range of 0.032-4.0μg/mL; 2. Immobilized human Leptin at 5μg/mL (100μL/well) can bind human Leptin receptor-His with a linear range of 0.032-4.0μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3952
≥95% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Lectins & Leptins
Leptin
Assay Standard
E. coli
Lyophilized
16kDa
5μg
Native
FLJ94114, OB, OBS
LEP
Other Proteins
Untagged
Documents

For Research Use Only. Not for use in diagnostic procedures.