Please login to your online account to display your discounted pricing

Sino Biological™ LDLR Recombinant Human Protein, His Tag DFS Item


This recombinant human protein is expressed from a DNA sequence encoding the extracellular domain of human LDLR precursor (NP_000518.1), residues 1 to 788, fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10231H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10231H08H50

  • / Each
 

Description & Specifications

Specifications

Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene Symbol LDLR
Quantity 50μg
Common Name Low density lipoprotein receptor
Validated Application Assay Standard
Expression System Mammalian
Endotoxin Concentration <1 EU/μg
Gene Alias FH, FHC, LDLCQ2, LDLR
Protein Family Other Proteins
Format Lyophilized
Purity ≥85% by SDS-PAGE
Gene ID (Entrez) 3949
Structural Form Truncated/Recombinant
Protein Tag His Tag
Molecular Weight 86kDa
Protein Subtype Other Proteins

Low density lipoprotein receptor (LDLR) recombinant human protein is supplied as a lyophilized powder. This protein is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.&line-break;Background: Low-density lipoprotein (LDL) is one of the five major groups of lipoprotein that transports cholesterol and triglycerides from the liver to peripheral tissues, and enables fats and cholesterol to move within the water-based solution of the blood stream. The Low-Density Lipoprotein Receptor (LDLR) is the prototype member of the LDLR family of scavenger receptors which is involved in receptor-mediated endocytosis of specific ligands. In order to be internalized, the receptor-ligand complexes must first cluster into clathrin-coated pits. LDLR recognizes the apoprotein B100 which is embedded in the phospholipid outer layer of LDL particles, as well as the apoE protein found in chylomicron remnants and VLDL remnants (IDL). After taken into the cell ending up in lysosomes, LDL is degraded and the cholesterol is made available for repression of microsomal enzyme HMG CoA reductase, the rate-limiting step in cholesterol synthesis. As a mosaic protein, the extracellular region of LDLR consists of seven N-terminal complement-like cysteinerich repeats followed by three EGF like repeats with a β-propeller YWTD containing motif, and a membrane proximal Ser/Thr rich region. Furthermore, it is reported that in case of HIV-1 infection, LDLR functions as a receptor for extracellular Tat (HIV-1 transactivator) into neurons, mediating its internalization in uninfected cells. Mutations in LDLR gene cause the autosomal dominant disorder, familial hypercholesterolemia.

  • N-terminal Sequence Analysis: Ala 22
  • Activity: Measure by its ability to bind to human PCSK9 in a functional ELISA: 1. Immobilized human PCSK9 at 10μg/mL (100μL/well) has been shown to bind biotinylated recombinant human LDLR, the ED50 of biotinylated human LDLR is 0.61μg/mL; 2. Immobilized mouse PCSK9 at 10μg/mL (100μL/well) has been shown to bind biotinylated recombinant human LDLR, the ED50 of biotinylated human LDLR is 0.12μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% mannitol, 5% trehalose, and 0.02% Tween®-80
  • Reconstitution: Dissolve the protein in sterile double-distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Manufactured by Sino Biological (China).