Please login to your online account to display your discounted pricing

Sino Biological™ LCN2 Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse LCN2 precursor (NP_032517.1) (Met 1-Asn 200) fused to a polyhistidine tag at the C-terminus

$96.50 - $1,320.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Lcn2, RP23-161B9.11, 24p3, AW212229, Sip24
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50060M08H25

DFS Item
 
life technologies
LS50060M08H25
5 x 5μg Each for $210.00

50060M08H5

DFS Item
 
life technologies
LS50060M08H5
5μg Each for $96.50

50060M08H50

DFS Item
 
life technologies
LS50060M08H50
50μg Each for $378.00

50060M08HL

DFS Item
 
life technologies
LS50060M08H250
5 x 50μg Each for $1,320.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Lcn2, RP23-161B9.11, 24p3, AW212229, Sip24

Lipocalin 2 (LCN2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing functional assays. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind Iron(III) dihydroxybenzoic acid (Fe(DHBA)3); the binding of Fe(DHBA)3 results in the quenching of Trp fluorescence in recombinant mouse Lipocalin 2 (LCN2); recombinant mouse LCN2 can bind >1.0μM of Fe(DHBA)3
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.