Please login to your online account to display your discounted pricing

Sino Biological™ LCN2 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human Lipocalin 2 (LCN2) precursor (NP_005555.2) (Met 1-Gly 198) fused to the polyhistidine tag at the C-terminus

$97.75 - $1,320.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias NGAL, LCN2
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10222H08H25

DFS Item
 
life technologies
LS10222H08H25
5 x 5μg Each for $215.00

10222H08H5

DFS Item
 
life technologies
LS10222H08H5
5μg Each for $97.75

10222H08HL

DFS Item
 
life technologies
LS10222H08H250
5 x 50μg Each for $1,320.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias NGAL, LCN2

Lipocalin 2 (LCN2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind Iron(III) dihydroxybenzoic acid (Fe(DHBA)3); the binding of Fe(DHBA)3 results in the quenching of Trp fluorescence in recombinant human Lipocalin-2
    Recombinant human Lipocalin-2 can bind >1.5μM of Fe(DHBA)3
  • Formulation: Lyophilized in 25mM MES, 150mM NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.