Please login to your online account to display your discounted pricing

Sino Biological™ LCN2 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human Lipocalin 2 (LCN2) precursor (NP_005555.2) (Met 1-Gly 198) fused to the polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10222H08H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10222H08HL

  • / Each
 

Description & Specifications

Specifications

Molecular Weight 22kDa
Purity ≥90% by SDS-PAGE
Quantity 5 x 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Subtype Other Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias NGAL, LCN2
Common Name Lipocalin 2
Gene Symbol LCN2
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized

Lipocalin 2 (LCN2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to bind Iron(III) dihydroxybenzoic acid (Fe(DHBA)3); the binding of Fe(DHBA)3 results in the quenching of Trp fluorescence in recombinant human Lipocalin-2
    Recombinant human Lipocalin-2 can bind >1.5μM of Fe(DHBA)3
  • Formulation: Lyophilized in 25mM MES, 150mM NaCl, pH 6.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.