Please login to your online account to display your discounted pricing

Sino Biological™ LAIR2 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human LAIR2 (NP_002279.2) (Met 1-Pro 152)

$205.00 - $205.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias LAIR2, XXbac-BCX535A19.6, CD306, mgC71634
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10868HNAH25

DFS Item
 
life technologies
LS10868HNAH25
5 x 5μg Each for $205.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias LAIR2, XXbac-BCX535A19.6, CD306, mgC71634

Leukocyte-Associated Immunoglobulin-Like Receptor 2 (LAIR2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.

  • Activity: Measured by the ability of the immobilized protein to support the adhesion of HT-29 human colon adenocarcinoma cells; when 5 x 104 cells/well are added to recombiannt human LAIR2 coated plates (50μg/mL with 100μL/well), approximately 30% will adhere after 30 minutes at 37°C (optimal concentration depends on cell type as well as the application or research objectives)
  • Formulation: Lyophilized in 50mM Tris, 0.7M NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.