Please login to your online account to display your discounted pricing

Sino Biological™ KNG1 Recombinant Mouse Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse KNG1 isoform 3 (NP_001095882.1) (Glu 21-Ser 480) fused to a polyhistidine tag at the C-terminus and a signal peptide at the N-terminus

$238.00 - $4,235.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Kng, mgC123437, Kng1
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50474M08H25

DFS Item
 
life technologies
LS50474M08H25
5 x 5μg Each for $700.00

50474M08H5

DFS Item
 
life technologies
LS50474M08H5
5μg Each for $238.00

50474M08H50

DFS Item
 
life technologies
LS50474M08H50
50μg Each for $1,182.00

50474M08HL

DFS Item
 
life technologies
LS50474M08H250
5 x 50μg Each for $4,235.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Kng, mgC123437, Kng1

Kininogen 1 (KNG1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to inhibit papain cleavage of a fluorogenic peptide substrate Z-FR-AMC, the IC50 value is <5nM
  • Formulation: Lyophilized in 20mM Tris, 100mM NaCl, pH 7.5, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.