Please login to your online account to display your discounted pricing

Sino Biological™ KLK8 Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the human KLK8 isoform 1 (O60259-1) (Met 1-Gly 260) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11820H08H50

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11820H08H50

  • / Each
 

Description & Specifications

Specifications

Common Name Kallikrein-related peptidase 8
Molecular Weight 26.4kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Validated Application Assay Standard
Format Lyophilized
Gene Alias UNQ283/PRO322, HNP, NP, NRPN, PRSS19, TADG14
Expression System Mammalian
Protein Family Other Proteins
Protein Subtype Proteases
Protein Tag His Tag
Quantity 50μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Gene Symbol KLK8

Kallikrein-related peptidase 8 (KLK8), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to inhibit thrombin cleavage of a fluorogenic peptide substrate Boc-VPR-AMC; the specific activity is >500pmole/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.