Please login to your online account to display your discounted pricing

Sino Biological™ KLK13 Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the pro-form of human KLK13 (NP_056411.1) (Met 1-Ile 262) fused to the polyhistidine tag at the C-terminus

$241.00 - $4,235.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias KLK13, DKFZp586J1923, KLK-L4, KLKL4
View More Specs

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

10199H08H25

DFS Item
 
life technologies
LS10199H08H25
5 x 5μg Each for $708.00

10199H08H5

DFS Item
 
life technologies
LS10199H08H5
5μg Each for $241.00

10199H08HL

DFS Item
 
life technologies
LS10199H08H250
5 x 50μg Each for $4,235.00

10199H08H50

DFS Item
 
life technologies
LS10199H08H50
50μg Each for $1,182.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias KLK13, DKFZp586J1923, KLK-L4, KLKL4

Kallikrein-related peptidase 13 (KLK13), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its ability to cleave the fluorogenic peptide substrate Boc-VPR-AMC; the specific activity is >500 pmoles/min./μg
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.