missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ KITLG Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 189) of human KITLG (P21583-1) fused to a polyhistidine tag at the C-terminus

$556.00 - $556.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias KITLG, DKFZp686F2250, FPH2, KL-1, Kitl, mgF, SCF, SF, SHEP7
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10451H08B50 Life Technologies
LS10451H08B50
50μg Each for $556.00
Specifications

Description

KIT ligand (KITLG/SCF), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a cell proliferation assay using TF-1 human erythroleukemic cells (Kitamura,T. et al. 1989, J. Cell Physiol. 140:323.), the EC50 for this effect is typically 0.2-1ng/mL
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
KITLG, DKFZp686F2250, FPH2, KL-1, Kitl, mgF, SCF, SF, SHEP7