missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ KITLG Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Ala 189) of human KITLG (P21583-1) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS10451H08B50

 View more versions of this product

Catalog No. 10451H08B50


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; KITLG Recombinant Human Protein, His Tag; 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

KIT ligand (KITLG/SCF), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured in a cell proliferation assay using TF-1 human erythroleukemic cells (Kitamura,T. et al. 1989, J. Cell Physiol. 140:323.), the EC50 for this effect is typically 0.2-1ng/mL
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
19.9kDa
50μg
Truncated/Recombinant
<1 EU/μg
KIT ligand
Assay Standard
Insect
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Growth Factors
KITLG, DKFZp686F2250, FPH2, KL-1, Kitl, mgF, SCF, SF, SHEP7
KITLG
Growth Factors & Receptors
His Tag