missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

KITL Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse KITL (P20826-1) (Met 1-Ala 189) fused to a polyhistidine tag at the C-terminus

$626.00 - $626.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias Clo, Con, Gb, Kitlg, mgf, SCF, SF, SLF, Sl, Steel, c ontrasted
View More Specs

Includes: Five vials of lyophilized protein

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50487M08B50 View Documents Life Technologies
LS50487M08B50
50μg Each for $626.00
 
Description

Description

KIT ligand (KITL), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: 1. Measured by its binding ability in a functional ELISA; immobilized mouse KITL at 2μg/mL (100μL/well) can bind mouse KIT with a linear ranger of 1.28-32ng/mL; 2. Measured in a cell proliferation assay using TF-1 human erythroleukemic cells (Kitamura, T. et al. 1989, J. Cell Physiol. 140:323), the EC50 for this effect is typically 0.5-5ng/mL
  • Formulation: Lyophilized in 20mM Tris, 500mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
<1 EU/μg
Lyophilized
Clo, Con, Gb, Kitlg, mgf, SCF, SF, SLF, Sl, Steel, c ontrasted

For Research Use Only. Not for use in diagnostic procedures.