Please login to your online account to display your discounted pricing

Sino Biological™ JAM2 Recombinant Mouse Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mouse JAM2 (NP_076333.3) extracellular domain (Met 1-Asn 236) fused to the Fc region of human IgG1 at the C-terminus

$205.00 - $205.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP23-253E17.2, 1110002N23Rik, 2410030G21Rik, 2410167M24Rik, AU016127, JAM-2, JAM-B
View More Specs
Includes:  Five vials of lyophilized protein

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

50464M02H25

DFS Item
 
life technologies
LS50464M02H25
5 x 5μg Each for $205.00
Description & Specifications

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias RP23-253E17.2, 1110002N23Rik, 2410030G21Rik, 2410167M24Rik, AU016127, JAM-2, JAM-B

Junction Adhesion Molecule 2 (JAM2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in performing cell based assays. It can also be used as a protein standard, or in other research applications.

  • Activity: Measured by the ability of the immobilized protein to support the adhesion of Jurkat human leukemic T cells; when 8 x 104 cells/well are added to JAM2 coated plates (0.2μg/mL and 100μL/well), approximately 40-55% will adhere specifically after 60 minutes at 37°C
  • Formulation: Lyophilized in 40mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.