missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ JAG1 Recombinant Human Protein, hIgG1-Fc Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the human JAG1 (NP_000205.1) extracellular domain (Met 1-Ser 1046) fused to Fc region of human IgG1 at the C-terminus

$97.75 - $1,322.00

Specifications

Purity >85% by SDS-PAGE
Purification Method PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias JAG1, AGS, AHD, AWS, CD339, HJ1, JAGL1, mgC104644
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
11648H02H25 Life Technologies
LS11648H02H25
5 x 5μg Each for $215.00
11648H02H5 Life Technologies
LS11648H02H5
5μg Each for $97.75
11648H02HL Life Technologies
LS11648H02H250
5 x 50μg Each for $1,322.00
Specifications

Description

Jagged 1 (JAG1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

>85% by SDS-PAGE
<1 EU/μg
JAG1, AGS, AHD, AWS, CD339, HJ1, JAGL1, mgC104644
PAGE
Lyophilized
182