missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ ISG15 Recombinant Human Protein DFS Item

This recombinant protein was expressed from a DNA sequence encoding the mature form of human ISG15 (AAH09507.1) (Met 1-Gly157)

Manufacturer:  Life Technologies LS12729HNAE5

 View more versions of this product

Catalog No. 12729HNAE5


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

ISG15 Ubiquitin-Like Modifier (ISG15), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, pH 8.0, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

17.9kDa
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
ISG15
Other Cytokines
G1P2, IFI15, IP17, UCRP, hUCRP
E. coli
<1 EU/μg
Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
ISG15 ubiquitin-like modifier
9636
Lyophilized
≥90% by SDS-PAGE
Other Proteins
5μg
Full Length/Recombinant
PAGE
Assay Standard