Please login to your online account to display your discounted pricing

Sino Biological™ INSR, Short Isoform, Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 944) of human INSR short isoform (NP_001073285.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11086H08H25

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11086H08H25

  • / Each
 

Description & Specifications

Specifications

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Gene ID (Entrez) 3643
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Purity ≥90% by SDS-PAGE
Protein Family Hormones & Receptors
Format Lyophilized
Endotoxin Concentration <1 EU/μg
Expression System Mammalian
Validated Application Assay Standard
KinaseFamily Insulin & IGF Receptor Kinase Family
Molecular Weight 106.0kDa
Quantity 5 x 5μg
Structural Form Truncated/Recombinant/Cell Surface Protein
Gene Alias CD220, HHF5, INSR
Common Name Insulin receptor, short isoform
Protein Tag His Tag
Protein Subtype Hormone Receptors
Gene Symbol INSR

Insulin receptor (INSR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; when 1μg/mL of biotinylated human insulin is immobilized onto a streptavidin coated plate, it can bind human insulin receptor (INSR) with a linear range of 0.3-40μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 25mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.