Please login to your online account to display your discounted pricing

Sino Biological™ INSR Recombinant Human Protein (Long Isoform, without Catalytic Activity Domain), His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Lys 956) of human INSR long isoform (NP_000199.2) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS11081H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 11081H08H5

  • / Each
 

Description & Specifications

Specifications

Format Lyophilized
Gene ID (Entrez) 3643
Structural Form Truncated/Recombinant/Cell Surface Protein
Common Name Insulin receptor, long isoform
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Purity ≥85% by SDS-PAGE
Validated Application Assay Standard
Molecular Weight 107.0kDa
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Quantity 5μg
Gene Alias CD220, HHF5, INSR
KinaseFamily Insulin & IGF Receptor Kinase Family
Expression System Mammalian
Endotoxin Concentration <1 EU/μg
Protein Tag His Tag
Protein Family Growth Factors & Receptors
Protein Subtype Hormone Receptors
Gene Symbol INSR

Insulin receptor (INSR), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications. Since this product is a truncated protein and does not contain intracellular catalytic activity domain, it is not suitable for use in enzyme activity studies.

  • Activity: Measured by its binding ability in a functional ELISA; when 1μg/mL of biotinylated human insulin is immobilized onto a streptavidin coated plate, it can bind human INSR, isoform long, with a linear range of 0.625-10μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween-80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.