missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL7R (CD127) Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 236) of human IL7R (NP_002176.2) fused to a polyhistidine tag at the C-terminus

$122.00 - $383.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IL7R, CD127, CDW127, IL-7R-alpha, IL7RA, ILRA
Gene ID (Entrez) 3575
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10975H08H25 View Documents Life Technologies
LS10975H08H25
5 x 5μg Each for $261.00
 
10975H08H5 View Documents Life Technologies
LS10975H08H5
5μg Each for $122.00
 
10975H08H50 View Documents Life Technologies
LS10975H08H50
50μg Each for $383.00
 
Description

Description

Interleukin 7 receptor (IL7R/CD127), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein-protein interaction assays, protein structure analysis, as immunogen, protein standard, and other research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL7R-His at 1μg/well can bind biotinylated mouse TSLP-His with a linear range of 1.25-10.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
3575
<1 EU/μg
IL7R, CD127, CDW127, IL-7R-alpha, IL7RA, ILRA

For Research Use Only. Not for use in diagnostic procedures.