Please login to your online account to display your discounted pricing

Sino Biological™ IL7R (CD127) Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 236) of human IL7R (NP_002176.2) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10975H08H5

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10975H08H5

  • / Each
 

Description & Specifications

Specifications

Gene Alias IL7R, CD127, CDW127, IL-7R-alpha, IL7RA, ILRA
Quantity 5μg
Molecular Weight 26.3kDa
Format Lyophilized
Expression System Mammalian
Common Name Interleukin 7 receptor
Protein Family Cytokines & Receptors/CD Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Truncated/Recombinant
Validated Application Assay Standard
Gene Symbol IL7R
Purity ≥90% by SDS-PAGE
Gene ID (Entrez) 3575
Protein Tag His Tag
Endotoxin Concentration <1 EU/μg
Protein Subtype Interleukins|Cytokine Receptors

Interleukin 7 receptor (IL7R/CD127), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein-protein interaction assays, protein structure analysis, as immunogen, protein standard, and other research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL7R-His at 1μg/well can bind biotinylated mouse TSLP-His with a linear range of 1.25-10.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.