missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IL7R (CD127) Recombinant Human Protein, His Tag DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gly 236) of human IL7R (NP_002176.2) fused to a polyhistidine tag at the C-terminus

Manufacturer:  Life Technologies LS10975H08H250

 View more versions of this product

Catalog No. 10975H08HL


 

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Specifications

Description

Protein; Sino Biological; IL7R (CD127) Recombinant Human Protein, His Tag; 5 x 50ug; Source: Human; Purity: greater than or equal to 90% by SDS-PAGE

Interleukin 7 receptor (IL7R/CD127), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein-protein interaction assays, protein structure analysis, as immunogen, protein standard, and other research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized human IL7R-His at 1μg/well can bind biotinylated mouse TSLP-His with a linear range of 1.25-10.0μg/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
3575
≥90% by SDS-PAGE
Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Interleukins|Cytokine Receptors
IL7R, CD127, CDW127, IL-7R-alpha, IL7RA, ILRA
IL7R
Cytokines & Receptors/CD Proteins
His Tag
Lyophilized
26.3kDa
5 x 50μg
Truncated/Recombinant
<1 EU/μg
Interleukin 7 receptor
Assay Standard
Mammalian