missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sino Biological™ IL6ST Recombinant Mouse Protein, hIgG1-Fc.His Tag, CHO expressed DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Glu 617) of mouse IL6ST (NP_034690.3) fused to the polyhistidine-tagged Fc region of human IgG1 at the C-terminus

$91.00 - $802.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IL6ST, 5133400A03Rik, AA389424, BB405851, CD130, gp130
Gene ID (Entrez) 16195
View More Specs

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
50135M03C25 Life Technologies
LS50135M03C25
5 x 5μg Each for $232.00
50135M03C5 Life Technologies
LS50135M03C5
5μg Each for $91.00
50135M03CL Life Technologies
LS50135M03C250
5 x 50μg Each for $802.00
Specifications

Description

Interleukin 6 signal transducer (IL6ST), recombinant mouse protein is supplied as a lyophilized powder. In general, recombinant proteins can be used as protein standard and in cell biology research applications.
  • Activity: Measured by its ability to inhibit the IL6R enhancement of IL6 activity on M1 mouse myeloid leukemia cells (Saito, T. et al. 1991, J. Immunol. 147:168); the ED50 for this effect is typically 0.03-0.1μg/mL in the presence of 50ng/mL recombinant human IL6R and 100ng/mL recombinant human IL6
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Description & Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
16195
<1 EU/μg
IL6ST, 5133400A03Rik, AA389424, BB405851, CD130, gp130