missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

IL6R Recombinant Human Protein, His Tag, Invitrogen™ Sino Biological™ DFS Item

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 365) of human IL6R (NP_000556.1) fused to a polyhistidine tag at the C-terminus

$112.00 - $484.00

Specifications

Purity ≥90% by SDS-PAGE
Endotoxin Concentration <1 EU/μg
Format Lyophilized
Gene Alias IL6R, CD126, IL-6R-1, IL-6R-alpha, IL6RA, mgC104991
Gene ID (Entrez) 3570
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
10398H08H25 View Documents Life Technologies
LS10398H08H25
5 x 5μg Each for $290.00
 
10398H08H5 View Documents Life Technologies
LS10398H08H5
5μg Each for $112.00
 
10398H08H50 View Documents Life Technologies
LS10398H08H50
50μg Each for $484.00
 
Description

Description

Interleukin 6 receptor (IL6R), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human IL6 at 8μg/mL (100μL/well) can bind recombinant human IL6R with a linear range of 1.25-20.0ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

≥90% by SDS-PAGE
Lyophilized
3570
<1 EU/μg
IL6R, CD126, IL-6R-1, IL-6R-alpha, IL6RA, mgC104991

For Research Use Only. Not for use in diagnostic procedures.