Please login to your online account to display your discounted pricing

Sino Biological™ IL6R Recombinant Human Protein, His Tag DFS Item


This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Pro 365) of human IL6R (NP_000556.1) fused to a polyhistidine tag at the C-terminus

Manufacturer: life technologies  LS10398H08H250

 View more versions of this product

 Disclaimers

For Research Use Only. Not for use in diagnostic procedures.

Catalog No. 10398H08HL

  • / Each
 

Description & Specifications

Specifications

Gene ID (Entrez) 3570
Quantity 5 x 50μg
Purity ≥90% by SDS-PAGE
Protein Family Cytokines & Receptors/CD Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene Alias IL6R, CD126, IL-6R-1, IL-6R-alpha, IL6RA, mgC104991
Molecular Weight 40kDa
Common Name Interleukin 6 receptor
Structural Form Truncated/Recombinant
Gene Symbol IL6R
Expression System Mammalian
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Protein Tag His Tag
Endotoxin Concentration <1 EU/μg
Protein Subtype Interleukins|Cytokine Receptors
Validated Application Assay Standard

Interleukin 6 receptor (IL6R), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.

  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human IL6 at 8μg/mL (100μL/well) can bind recombinant human IL6R with a linear range of 1.25-20.0ng/mL
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.